Learn More
UBXN2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564224
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MKDVDNLKSI KEEWVCETGS DNQPLGNNQQ SNCEYFVDSL FEEAQKVSSK CVSPAEQKKQ VDVN Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
UBXN2A Polyclonal antibody specifically detects UBXN2A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
UBXN2A | |
Polyclonal | |
Unconjugated | |
UBXN2A | |
6330407P03Rik; UBX domain containing 4; UBX domain protein 2A; UBX domain-containing protein 2A; UBX domain-containing protein 4; Ubxd4; Ubxn2a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
165324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P68543 | |
UBXN2A | |
Recombinant protein corresponding to Human UBXN2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only