missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Rabbit Polyclonal Antibody, Epredia™
Ubiquitin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
£106.08 - £450.00
Specifications
Antigen | Ubiquitin |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12683467
|
Epredia
RB-9203-P1 |
Concentrated | 500 μL |
£337.00
500µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12673467
|
Epredia
RB-9203-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12623467
|
Epredia
RB-9203-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12603477
|
Epredia
RB-9203-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by a 26S complex, multicatalytic cytosolic and nuclear protease termed proteasome. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant full-length human ubiquitin protein
Molecular Weight: 8.5kDa
Positive Control: Human brain from patient with Alzheimer's disease
Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Ubiquitin | |
Polyclonal | |
Rabbit | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Recombinant full-length human ubiquitin protein | |
Primary |