missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBOX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567607
Additional Details : Weight : 0.01000kg
Description
UBOX5 Polyclonal antibody specifically detects UBOX5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
UBOX5 | |
Polyclonal | |
Unconjugated | |
Ubox5 | |
1500010O06Rik; C330018L13Rik; FAST kinase domain-containing protein 5; RING finger protein 37; FAST kinase domains 5; FASTKD5; hUIP5; KIAA0860; RGD1305440; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; Rnf37; U box domain containing 5; UBCE7IP5; ubcM4-interacting protein 5; Ube7ip5; ubiquitin conjugating enzyme 7 interacting protein 5; ubiquitin-conjugating enzyme 7-interacting protein 5; U-box domain containing 5; U-box domain-containing protein 5; Ubox5; UIP5; Unknown (protein for MGC:138103) | |
Rabbit | |
affinity chromatography | |
RUO | |
140629, 22888 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O94941, Q925F4 | |
Ubox5 | |
Fusion protein of human UBCE7IP5 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |