missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25122-1-AP-150UL
This item is not returnable.
View return policy
Specifications
UBAC2 | |
Polyclonal | |
Unconjugated | |
UBAC2 | |
PHGDHL1, UBA domain containing 2, UBAC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
337867, 68889 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NBM4, Q8R1K1 | |
UBAC2 | |
UBAC2 Fusion Protein Ag19142 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |