Learn More
UAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553206
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NDLKLTLSKA GQEHLLRFWN ELEEAQQVEL YAELQAMNFE ELNFFFQKAI EGFNQSSHQK NVDARMEPVP REVLGSATRD QDQLQAWESE GLFQISQNKV AVLLLAGGQG TRLGVAYPKG MYDVGLPSRK TLFQIQAERI LKLQQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.
UAP1 Polyclonal antibody specifically detects UAP1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
UAP1 | |
Polyclonal | |
Unconjugated | |
UAP1 | |
AA420407; AA437972; AgX; AGX1; AGX-1; AGX2; AGX-2; antigen X; ESTM38; fb63d12; RGD1561967; similar to UDP-N-acetylhexosamine pyrophosphorylase (Antigen X) (AGX) (Sperm-associated antigen 2); SPAG2; sperm associated antigen 2; sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UAP1; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylglucosamine pyrophosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1; UDP-N-acteylglucosamine pyrophosphorylase 1; wu:fb63d12; zgc:55280 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16222 | |
UAP1 | |
Recombinant protein corresponding to Human UAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only