missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers, Model IC 458
Made of high density polyethylene, the Tyvek™ IsoClean™ Boot Covers provide an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
215.00€
Specifications
Material | HDPE |
---|---|
Color | White |
Sterility | Non-sterile |
Closure Type | Ties |
Sole Type | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16468353
|
DuPont™
D15531626 |
Medium |
215.00€
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
16478353
|
DuPont™
D15531627 |
Large |
215.00€
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
- Gripper™ soles provide enhanced slip-retardance and robustness.
- Delivers an ideal balance of protection, durability and comfort.
- Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
Specifications
HDPE | |
Non-sterile | |
Slip-resistant | |
Serged | |
Boot Covers |
White | |
Ties | |
IC 458 | |
(EU) 2016/425, EN 14126 |