Learn More
S.C.A.T™ Blind Plug
SCAT ™ blind plugs close unneded ports of SafetyCaps and SafetyWasteCaps.
£14.38 - £39.42
Specifications
Material PTFE
View More Specs
Products 2
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
13233859
View Documents
S.C.A.T™
160501
SCAT SafetyCaps and SafetyWasteCaps £39.42
Pack of 10
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 2
Add to basket
10051453
View Documents
S.C.A.T™
160506
SCAT SafetyCaps and SafetyWasteCaps
£14.38
Each
View alternative products
Save Up to 
»
Estimated Shipment
06-07-2021
Add to basket
Description
Description
  • Blind plugs for the S.C.A.T™ Europe Safety System
  • Made from PFA or PTFE
  • Resistant against aggressive media
  • Blind plugs close unneded ports
  • Can close capillary ports, tube ports and exhaust filter ports
Specifications
Specifications
PTFE
SDS