missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%, Thermo Scientific Chemicals

92.20 EUR - 336.00 EUR

Chemical Identifiers

CAS 15492-52-1
Molecular Formula C33H57O6Yb
Molecular Weight (g/mol) 722.867
MDL Number MFCD00000034
InChI Key KACMHCBXMZUMQV-LWTKGLMZSA-K
Synonym 2,2,6,6-tetramethylheptane-3,5-dione; ytterbium 3+, ytterbium 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato ytterbium iii-yb reo yb tmhd 3
PubChem CID 44473428
IUPAC Name (Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;ytterbium(3+)
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Yb+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374487
View Documents
Thermo Scientific Alfa Aesar
039581.03
1 g
92.20 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
11384487
View Documents
Thermo Scientific Alfa Aesar
039581.06
5 g
336.00 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15492-52-1
722.867
KACMHCBXMZUMQV-LWTKGLMZSA-K
44473428
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Yb+3]
C33H57O6Yb
MFCD00000034
2,2,6,6-tetramethylheptane-3,5-dione; ytterbium 3+, ytterbium 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato ytterbium iii-yb reo yb tmhd 3
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;ytterbium(3+)

Specifications

15492-52-1
C33H57O6Yb
Hygroscopic
Insoluble in water.
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Yb+3]
722.867
722.86
Crystalline
167°C to 169°C
MFCD00000034
2,2,6,6-tetramethylheptane-3,5-dione; ytterbium 3+, ytterbium 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato ytterbium iii-yb reo yb tmhd 3
KACMHCBXMZUMQV-LWTKGLMZSA-K
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;ytterbium(3+)
44473428
99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only