missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number p-4510527
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 mL
10 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
1493-13-6
CHF3O3S
150.07
MFCD00007514
ITMCEJHCFYSIIV-UHFFFAOYSA-N
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
62406
CHEBI:48511
OS(=O)(=O)C(F)(F)F
1493-13-6
CHF3O3S
150.07
MFCD00007514
ITMCEJHCFYSIIV-UHFFFAOYSA-N
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
62406
CHEBI:48511
OS(=O)(=O)C(F)(F)F

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.6900g/mL
Boiling Point 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3SO3H
Refractive Index 1.3260 to 1.3300
Quantity 1 mL
Beilstein 03,IV,34
Fieser 04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24
Merck Index 15,9842
Specific Gravity 1.69
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an
IUPAC Name trifluoromethanesulfonic acid
Viscosity 2.87 mPa.s (25°C)
Formula Weight 150.07
Percent Purity 99%
Grade Extra Pure
Physical Form Fuming Liquid
Chemical Name or Material Trifluoromethanesulfonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoromethanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'