missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 98%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

£62.91 - £186.93

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398865
View Documents
Thermo Scientific Alfa Aesar
L13994.14
25 g
£62.91
25g
Please to purchase this item. Need a web account? Register with us today!
11308875
View Documents
Thermo Scientific Alfa Aesar
L13994.22
100 g
£186.93
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and chloroform. Immiscible with water.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

156-60-5
96.94
KFUSEUYYWQURPO-OWOJBTEDSA-N
638186
(E)-1,2-dichloroethene
C2H2Cl2
MFCD00062942
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
CHEBI:29027
Cl\C=C\Cl

Specifications

156-60-5
1.252
2°C (35°F)
1.4455
UN1150
14,92
Miscible with ethanol,ethyl ether,acetone,benzene and chloroform. Immiscible with water.
Cl\C=C\Cl
96.94
CHEBI:29027
98%
-50°C
47°C to 49°C
C2H2Cl2
MFCD00062942
1420761
trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
KFUSEUYYWQURPO-OWOJBTEDSA-N
(E)-1,2-dichloroethene
638186
96.94
trans-1,2-Dichloroethylene, stabilized with 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P372-P374-P380-P501c

H225-H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DICHLOROETHYLENE

missing translation for 'einecsNumber' : 205-860-2

missing translation for 'rtecsNumber' : KV9400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only