missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMUB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | TMUB2 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16805975
|
Proteintech
28044-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16895965
|
Proteintech
28044-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMUB2 (Transmembrane And Ubiquitin Like Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is TMUB1.Specifications
TMUB2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q71RG4 | |
79089 | |
TMUB2 Fusion Protein Ag27691 | |
Primary | |
-20°C | |
TMUB2 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FP2653, TMUB2, UNQ1897/PRO4343 | |
TMUB2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |