missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMUB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28044-1-AP-20UL
This item is not returnable.
View return policy
Description
TMUB2 (Transmembrane And Ubiquitin Like Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is TMUB1.Specifications
TMUB2 | |
Polyclonal | |
Unconjugated | |
TMUB2 | |
FP2653, TMUB2, UNQ1897/PRO4343 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79089 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q71RG4 | |
TMUB2 | |
TMUB2 Fusion Protein Ag27691 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |