Learn More
Invitrogen™ Thyroid Peroxidase Human ELISA Kit
Sandwich ELISA Kit
£504.00 - £2320.00
Specifications
Accession Number | P07202 |
---|---|
Assay Sensitivity | 0.3 ng/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485163
|
Invitrogen™
EHTPO |
96 Tests |
£504.00
1 set |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17125599
|
Invitrogen™
EHTPOX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
The Human Thyroid Peroxidase ELISA quantitates Hu Thyroid Peroxidase in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Thyroid Peroxidase. Principle of the method The Human Thyroid Peroxidase solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes a membrane-bound glycoprotein. The encoded protein acts as an enzyme and plays a central role in thyroid gland function. The protein functions in the iodination of tyrosine residues in thyroglobulin and phenoxy-ester formation between pairs of iodinated tyrosines to generate the thyroid hormones, thyroxine and triiodothyronine. Mutations in this gene are associated with several disorders of thyroid hormonogenesis, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional splice variants have been described but their biological natures have not been determined.Specifications
P07202 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
MSA,TDH2A,TPX | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.3 ng/mL | |
Biotin | |
Proteases, Peptidases & Inhibitors | |
This ELISA pair antibody detects human TPX. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
7173 | |
<12% | |
HRP | |
Endocrinology | |
MSA, TDH2A, TPX | |
1 hr. 20 min. |