Learn More
Thermo Scientific™ SWAP-IT No Vent System
Easy to install for quick column changeovers
£38.17
Products 1
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
11717436
View Documents
Thermo Scientific™
290SG020
Graphite/Vespel Ferrule; For Thermo Scientific Transfer Line 10/Pk
£38.17
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 25-08-2021
Add to basket
Description
Description
The Thermo Scientific SWAP-IT GCMS Interface provides a powerful tool for switching capillary columns without the need to vent the MS.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution