missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters - 25mm Diameter

Accommodate sample volumes of 10 to 50mL

£119.65 - £216.14


Products 8
Product Code Brand For Use With (Application) Material No. per Pack Pore Size Price Quantity & Availability  
Product Code Brand For Use With (Application) Material No. per Pack Pore Size Price Quantity & Availability  
10302961
View Documents
Thermo Scientific™
191-2080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10578091
View Documents
Thermo Scientific™
196-2020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10508481
View Documents
Thermo Scientific™
199-2020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10730622
View Documents
Thermo Scientific™
196-2045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10603711
View Documents
Thermo Scientific™
199-2045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10692741
View Documents
Thermo Scientific™
191-2020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10682292
View Documents
Thermo Scientific™
194-2545
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10162351
View Documents
Thermo Scientific™
194-2520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding
  • Ideal for sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids
  • Have no wetting agents
  • Modified acrylic housings
  • Membrane of choice for sterilization
Cellulose Acetate (CA)
  • Low protein binding
  • Ideal for sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Modified acrylic housings
  • Sterile
Nylon
  • Naturally hydrophilic, low extractables
  • Useful for sterilizing cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC solvent applications
  • Polypropylene housings
  • Autoclavable

SDS
Product Certifications