Learn More
Amarell™ Thermo-Hygrometer With Outdoor Probe
Use 3m cable for outdoor readings. The Thermo-Hygrometer provides readings between -50 to 70°C and 20 to 90% rF.
Brand: Amarell™ E915000
This item is not returnable.
View return policy
Description
Universal thermo-hygrometer with outdoor probe.- Accessory for wall mounting (screws or adhesive tape).
- Indoor/Outdoor
- Min/Max Function
- °C ↔ °F
- With Clock
- Fast responding semi-conductor probe
- Probe includes 3m cable for outdoor measurements
Specifications
Thermohygrometer | |
1% rF (rh) | |
-50°C to +70°C | |
0.1°C | |
3 m | |
3m cable and probe |
20 to 90% | |
±5% rF (rh) | |
±1°C | |
110 x 60 x 20 mm | |
1.5 V |