missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep™
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Certificate of Analysis | Assurance Certificate of Analysis reports up to 30 elements at ppb levels. |
Safety and Handling
Product Identifier
- 1000 µg/mL Thallium
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : nitric acid, thallium nitrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'