Learn More
Tetramethoxysilane, 98%, Thermo Scientific Chemicals
£25.33 - £191.96
Chemical Identifiers
CAS | 681-84-5 |
---|---|
Molecular Formula | C4H12O4Si |
Molecular Weight (g/mol) | 152.22 |
MDL Number | MFCD00008341 |
InChI Key | LFQCEHFDDXELDD-UHFFFAOYSA-N |
Synonym | tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate |
PubChem CID | 12682 |
IUPAC Name | tetramethyl silicate |
SMILES | CO[Si](OC)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456760
|
Thermo Scientific Alfa Aesar
L09716.14 |
25 g |
£25.33
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466760
|
Thermo Scientific Alfa Aesar
L09716.22 |
100 g |
£59.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15468909
|
Thermo Scientific Alfa Aesar
L09716.36 |
500 g |
£191.96
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.
Solubility
Miscible with ethanol and organic solvents. Immiscible with water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.
Chemical Identifiers
681-84-5 | |
152.22 | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
12682 | |
CO[Si](OC)(OC)OC |
C4H12O4Si | |
MFCD00008341 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
tetramethyl silicate |
Specifications
121°C to 122°C | |
681-84-5 | |
MFCD00008341 | |
1699658 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
tetramethyl silicate | |
12682 | |
98% | |
1.368 | |
1.032 |
4°C to 5°C | |
C4H12O4Si | |
UN2606 | |
26°C (78°F) | |
Miscible with ethanol and organic solvents. Immiscible with water. | |
CO[Si](OC)(OC)OC | |
152.22 | |
152.22 | |
Ether-like | |
Moisture sensitive | |
Tetramethoxysilane |
Safety and Handling
GHS H Statement
H330-H318-H226-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ORTHOSILICATE
missing translation for 'einecsNumber' : 211-656-4
missing translation for 'rtecsNumber' : VV9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only