missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butyltin, 95%, Thermo Scientific Chemicals

Product Code. 11317717
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetra-n-butyltin is used in hydrogenolysis reactions and as a transition metal catalyst. It is also used stabilizers in the manufacture of polyvinyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-25-2
Molecular Formula C16H36Sn
Molecular Weight (g/mol) 347.174
MDL Number MFCD00009422
InChI Key AFCAKJKUYFLYFK-UHFFFAOYSA-N
Synonym tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
PubChem CID 15098
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Melting Point -97°C
Density 1.057 g/mL
Boiling Point 144°C to 146°C (10 mmHg)
Flash Point 107°C (225°F)
Odor Mild
Refractive Index 1.474
Quantity 500 g
UN Number UN2788
Solubility Information Insoluble in water. It is soluble in most common organic solvents.
Formula Weight 347.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'