missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, Puriss. p.a., ≥99.5% (GC), Honeywell Riedel-de Haën™
£51.39 - £102.73
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15661740
|
Honeywell-Riedel-de Haen
20256-1L |
1 L | Glass Bottle |
£51.39
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15671740
|
Honeywell-Riedel-de Haen
20256-2.5L |
2.5 L | Glass Bottle |
£102.73
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1634-04-4 | |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
Specifications
-109°C | |
0.74g/mL at 25°C, 0.736 to 0.744g/mL at 20°C | |
0.151 | |
705°F | |
C5H12O | |
MFCD00008812 | |
1730942 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.15g/mol | |
puriss. p.a. |
1634-04-4 | |
55.3°C | |
−33°C (−27.4°F) | |
1.376 | |
(CH3)3COCH3 | |
UN2398 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
≥99.5% (GC) | |
Tert-Butyl Methyl Ether |
Safety and Handling
P210
H225-H315
missing translation for 'dotInformation' : 3/PGII
missing translation for 'einecsNumber' : 216-653-1
missing translation for 'rtecsNumber' : KN5250000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- tert-Butyl methyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
Precautionary Statement
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.