Learn More
Techne Thermal cycler primeG gradient with touchscreen and up to 3.4°C/sec ramp rate 100-230V 50/60Hz 96mL x 0.2mL 420mm x 240mm x 240mm Techne
Brand:  Techne 5PRIMEG/02
Thermal cycler primeG gradient with touchscreen and up to 3.4°C/sec ramp rate 100-230V 50/60Hz 96mL x 0.2mL 420mm x 240mm x 240mm Techne
Code : 04
Additional Details : Weight : 9.40000kg
Product Code. 13521950