Learn More
Techne Thermal cycler prime with colour touchscreen and 3.0°C/sec ramp rate 100-230V 50/60Hz 155W 24mL x 0.2mL 350mm x 210mm x 180mm Techne
Brand:  Techne 3PRIMEBASE/02
Thermal cycler prime with colour touchscreen and 3.0°C/sec ramp rate 100-230V 50/60Hz 155W 24mL x 0.2mL 350mm x 210mm x 180mm Techne
Code : 04
Additional Details : Weight : 6.00000kg
Product Code. 13531940