Learn More
Micronova™ Tape Controlled Environment
PCX™ Cleanroom tape can be used for colour-coding, sealing, labelling and masking, and various tasks within the cleanroom requiring pressure-sensitive qualities
Brand:  Micronova™ PCX-1BL
Product Code. 13168485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
polyethylene, black 25mm x 33m Premium
Specifications
Specifications
Black
Polyethylene
Cleanroom use - color-coding, sealing, labelling and masking, or for pressure-sensitive applications
2.5 cm
Roll
Non-sterile
33 m