Learn More
Thermo Scientific™ SureQuant™ AKT Isoform Relative Quantitation Module
Thermo Scientific™ SureQuant™ AKT Isoform Relative Quantitation Module provides a complete quantitation solution for the analysis of key AKT expression using mass spectrometry (MS).
Brand: Thermo Scientific™ A40092
This item is not returnable.
View return policy
Description
- Complete - contains validated AQUA HeavyPeptide mix, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
- Validated - peptides rigorously tested for successful quantitation of each target peptide
- Multiplex - able to quantitate AKT1, AKT2, and AKT3 isoforms and phosphorylation of AKT
Specifications
Mass Spectrometry | |
Approved for shipment on Wet or Dry Ice | |
SureQuant™ | |
Mass Spectrometry | |
Peptides | |
10 Reactions |
Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer | |
Target Selection | |
1 Kit | |
Quantitation Module | |
Protein Samples |
For Research Use Only. Not for use in diagnostic procedures.