missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STP 7 Model Buffer Solution pH 7, WTW™
Click to view available options
Quantità:
50 mL
Imballaggio:
Bottle
Descrizione
- Technical buffering solutions for pH 7.00
- Precise and traceable to NIST in bulk size of 50 ml with built-in dosing vessel standard buffer
- Life up to 3 years (unopened)
- Thrifty use
- Easiest handling by means of dosing device
Specifica
Specifica
pH | 7 |
Forma fisica | Liquid |
Quantità | 50 mL |
Imballaggio | Bottle |
Tracciabilità a NIST | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'