Learn More
RUBIS Long Tail Tweezers
Long Tail Tweezers - Antimagnetic/Anti-acid SS, Serrated Handle
POA
Specifications
Tip Style | Straight, Round, Blunt, Serrated |
---|---|
Material | Anti-acid, Anti-magnetic Stainless Steel |
Product Type | Long Tail Tweezers |
Quantity (No. per Case) | Each |
Description
- Long Tail Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Serrated handles
- Serrated tips for better grip
- Blunt tips
- Suitable for cryogenics applications
- Strong
Specifications
Straight, Round, Blunt, Serrated | |
Anti-acid, Anti-magnetic Stainless Steel | |
Long Tail Tweezers | |
Each |