missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Stachyose hydrate, 97+%

£100.50 - £813.05

Chemical Identifiers

CAS 54261-98-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
MDL Number MFCD00149457
InChI Key UQZIYBXSHAGNOE-CYDMKDQUNA-N
PubChem CID 131801001
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10711311
View Documents
Thermo Scientific Chemicals
226085000
500 mg Glass bottle
£100.50
500mg
Please to purchase this item. Need a web account? Register with us today!
10701311
View Documents
Thermo Scientific Chemicals
226080010
1 g Glass bottle
£174.87
1g
Please to purchase this item. Need a web account? Register with us today!
10583694
View Documents
Thermo Scientific Chemicals
226080050
5 g Glass bottle
£813.05
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54261-98-2
666.58
UQZIYBXSHAGNOE-CYDMKDQUNA-N
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
C24H42O21
MFCD00149457
131801001

Specifications

Brown to Colorless
Liquid
Authentic
+129
MFCD00149457
UQZIYBXSHAGNOE-CYDMKDQUNA-N
2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate
131801001
≥97%
54261-98-2
95.0°C to 105.0°C
97% min. (ELSD) (HPLC)
C24H42O21
5% in water, Clear to slightly hazy, Colorless
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
666.58
666.58
Stachyose hydrate, 98.5%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Stachyose hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only