missing translation for 'onlineSavingsMsg'
Learn More

Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals

£65.40 - £195.98

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442057
View Documents
Thermo Scientific Alfa Aesar
044866.22
100 g
£65.40
100g
Please to purchase this item. Need a web account? Register with us today!
15452057
View Documents
Thermo Scientific Alfa Aesar
044866.36
500 g
£195.98
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Specifications

Gray
Dark gray sponge
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Air Sensitive
Nickel
7697-37-2
HNO3
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
Powder
Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing and gloves.

P201-P202-P210-P222-P235+P410-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P335+P334-P363-P370+P378q-P501c

H250-H317-H350-H372-X

missing translation for 'dotInformation' : DOT Class: 4.2, Packaging Group: II

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H251-Self-heating: may catch fire.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P422-Store contents under .
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Nickel, Aluminium, Chromium, Iron, Molybdenum

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only