missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | SPOCD1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851785
|
Proteintech
22243-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16841785
|
Proteintech
22243-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SPOCD1 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q6ZMY3 | |
90853 | |
SPOCD1 Fusion Protein Ag17734 | |
Primary | |
-20°C | |
SPOCD1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RP11 84A19.1, SPOC domain containing 1, SPOCD1 | |
SPOCD1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |