missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, Honeywell Fluka™

Product Code. 15605310
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 12 kg
Linear Formula NaI
UN Number UN3077
Formula Weight 149.89g/mol
Percent Purity ≥99.5%
Density Vapor Density: >1 (vs air)
Chemical Name or Material Sodium iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'