missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Product Code. 15603020
Click to view available options
Quantity:
100 g
250 g
1 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10039-56-2
Molecular Formula H4NaO3P
Molecular Weight (g/mol) 105.99
MDL Number MFCD06252379
InChI Key PLZNPHDJGFDNRM-UHFFFAOYSA-M
Synonym sodium hypophosphite hydrate, sodium phosphinate hydrate, sodium phosphenite hydrate, sodium hypophosphite en grade, sodium hydrate hypophosphite, sodium hypophosphite monohydrate 250g, phosphinic acid sodium salt monohydrate, sodium hypophosphite monohydrate, phosphonous acid monosodium salt monohydrate, sodium hypophosphite monohydrate, puriss
PubChem CID 23708894
IUPAC Name sodium hydrate phosphinate
SMILES O.[Na+].[O-][PH2]=O

Specifications

Quantity 100 g
Linear Formula NaH2PO2 · H2O
UN Number NONH for all modes of transport
Formula Weight 105.99g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Sodium Hypophosphite Monohydrate, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'