Learn More
Sodium formate, ACS, 99.0% min, Thermo Scientific Chemicals
Description
Reducing agent, buffering agent, complexing agent, Noble metal precipitantSodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
Melting Point | 259°C to 262°C |
Density | 1.92 g/mL |
Assay Percent Range | 0.99 |
Quantity | 2 kg |
Beilstein | 3595134 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water,glycerol. Slightly soluble in alcoholSoluble in water,alcohol,glycerol and formic acid. Insoluble in ether. |
Formula Weight | 68.01 |
Grade | ACS Reagent |
Physical Form | Crystalline |
Mehr anzeigen |
RUO – Research Use Only