missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichromate dihydrate, ACS, 99.5-100.5%, Thermo Scientific Chemicals

£26.73 - £203.01

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364217
View Documents
Thermo Scientific Alfa Aesar
036692.22
100 g
£26.73
100g
Please to purchase this item. Need a web account? Register with us today!
11354217
View Documents
Thermo Scientific Alfa Aesar
036692.36
500 g
£65.73
500g
Please to purchase this item. Need a web account? Register with us today!
11344217
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036692.A3
2 kg
£203.01
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-12-0
297.995
JYDRNIYTFCBIFC-UHFFFAOYSA-N
108005
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
Cr2H4Na2O9
MFCD00149166
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate

Specifications

100°C-2H2O
Red
99.5-100.5%
MFCD00149166
Soluble in water. Insoluble in alcohol.
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
297.995
298.00 (261.97 Anhydrous)
Hygroscopic
2.438 g/cm3 at 20°C
7789-12-0
Crystalline
Cr2H4Na2O9
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
JYDRNIYTFCBIFC-UHFFFAOYSA-N
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
108005
ACS
Red crystalline
Sodium dichromate dihydrate, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372

missing translation for 'dotInformation' : DOT Class: 5.1; Packing Group: II

missing translation for 'einecsNumber' : 234-190-3

missing translation for 'rtecsNumber' : HX7750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only