missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfate Monohydrate, 99%, ReagentPlus™, Honeywell™

Product Code. 15622320
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium hydrate hydrogen sulfate
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58°C
Color White
Quantity 1 kg
Linear Formula NaHSO4 · H2O
UN Number NONH for all modes of transport
Formula Weight 138.08g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Poly Bottle
Appearance Moist crystals
Density 2.12g/cm3
Chemical Name or Material Sodium Bisulfate Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'