missing translation for 'onlineSavingsMsg'
Learn More

CSF3R (CD114) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human CSF3R precursor (NP_000751.1) precursor fused to a polyhistidine tag at the C-terminus

POA

Specifications

Purity >85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD114, GCSFR, CSF3R
Gene ID (Entrez) 1441
View More Specs

Description

Description

Colony stimulating factor 3 receptor (CSF3R/CD114), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to inhibit the CSF3 induced proliferation of NFS60 mouse myeloid cells, the ED50 for this effect is typically 20-80ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>85% by SDS-PAGE
Lyophilized
1441
<1 EU/μg
CD114, GCSFR, CSF3R
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.