missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SH2D1A |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899223
|
Proteintech
11868-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889223
|
Proteintech
11868-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SAP kinase is induced by stress agonists such as UV irradiation, heat shock and inflammatory cytokines. These stress stimuli result in the phosphorylation of and enzymatic activation of Stress Activated Protein Kinases (SAP kinases). Three SAP kinase gene are known to exist, a, b and g , which are differentially spliced giving 10 gene products in total.Specifications
SH2D1A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
B2RZ59, O60880, O88890 | |
20400, 4068, 501502 | |
SH2D1A Fusion Protein Ag2450 | |
Primary | |
-20°C | |
SH2D1A |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DSHP, Duncan disease SH2 protein, EBVS, IMD5, LYP, MTCP1, SAP, SH2 domain protein 1A, SH2D1A, SLAM associated protein, XLP, XLPD | |
SH2D1A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |