missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11868-1-AP-150UL
This item is not returnable.
View return policy
Description
SAP kinase is induced by stress agonists such as UV irradiation, heat shock and inflammatory cytokines. These stress stimuli result in the phosphorylation of and enzymatic activation of Stress Activated Protein Kinases (SAP kinases). Three SAP kinase gene are known to exist, a, b and g , which are differentially spliced giving 10 gene products in total.Specifications
SH2D1A | |
Polyclonal | |
Unconjugated | |
SH2D1A | |
DSHP, Duncan disease SH2 protein, EBVS, IMD5, LYP, MTCP1, SAP, SH2 domain protein 1A, SH2D1A, SLAM associated protein, XLP, XLPD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20400, 4068, 501502 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B2RZ59, O60880, O88890 | |
SH2D1A | |
SH2D1A Fusion Protein Ag2450 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |