Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non-specific sample interaction and increased analyte recovery.
Brand: Trajan™ 002790
This item is not returnable.
View return policy
Description
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
Gas/Liquid chromatography | |
662°F | |
10R-RTC/RSH-GT-MS1 | |
10 μL | |
Removable |
Autosampler Syringe | |
350°C | |
Ea. | |
Thermo RSH and CTC RTC |