Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand: Trajan™ 003760
This item is not returnable.
View return policy
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Diamond MS Syringe | |
Ea. | |
Removable |
25R-AG-GT-MS1 | |
25 μL |