missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SAA2 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881284
|
Proteintech
13192-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871284
|
Proteintech
13192-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Major acute phase reactant. Apolipoprotein of the HDL complex.Specifications
SAA2 | |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P05367, P0DJI9 | |
20209, 6289 | |
SAA2 Fusion Protein Ag3869 | |
Primary | |
-20°C | |
SAA2 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SAA, SAA1, SAA2, Serum amyloid A protein, serum amyloid A2 | |
SAA2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |