missing translation for 'onlineSavingsMsg'
Learn More

RYR1 Mouse anti-Human, Mouse, Clone: 1B4E12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66539-1-IG-20UL

 View more versions of this product

Product Code. 16888505

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscled cells. Ryanodine receptor, ∽500 kDa, can be isolated from a variety of cell types such as lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscles, cardiac muscle and brain tissues.
Specifications

Specifications

RYR1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
E9PZQ0, P21817
RYR1
RYR1 Fusion Protein Ag25120
20 μL
Primary
Mouse, Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
1B4E12
Unconjugated
RYR1
CCO, MHS, MHS1, Ryanodine receptor 1, RYDR, RYR, RYR 1, RYR1, SKRR
Mouse
Protein G
RUO
20190, 6261
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers