missing translation for 'onlineSavingsMsg'
Learn More

Rubrene, 99%, Thermo Scientific Chemicals

Catalog Number p-191170
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
517-51-1
C42H28
532.67
MFCD00003703
YYMBJDOZVAITBP-UHFFFAOYSA-N
rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
68203
5,6,11,12-tetraphenyltetracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
This item is not returnable. View return policy
517-51-1
C42H28
532.67
MFCD00003703
YYMBJDOZVAITBP-UHFFFAOYSA-N
rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
68203
5,6,11,12-tetraphenyltetracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 330°C to 335°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 05, II, 726
Solubility Information Solubility in water: practically insoluble
Formula Weight 532.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Rubrene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'