Learn More
Bellingham Stanley™ Refractometer Chemical C2 (Ri 1.42/°C No Atc) Hand Held
Brand:  Bellingham Stanley™ 38-37
Code : 04
Additional Details : Weight : 0.10000kg
Product Code. 12918564
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
dual scale, supplied with 2 x 3V AAA batteries (LR03) 55mm x 120mm x 35mm (w x d x h) 1.33 RI to 1.42 RI OPTi