Learn More
Tenak™ Red Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand:  Tenak™ TE22502
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 15270594
  • £82.10 / Pack of 36
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Specifications
Red
Square
136 x 136 x 50 mm
Cardboard
Cryo Box