missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM8A/Y14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14958-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein with a conserved RNA-binding motif. The protein is found predomitly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.Specifications
RBM8A/Y14 | |
Polyclonal | |
Unconjugated | |
RBM8A | |
Binder of OVCA1 1, BOV 1, BOV 1A, BOV 1B, BOV 1C, MDS014, RBM8, RBM8A, RBM8B, Ribonucleoprotein RBM8A, RNA binding motif protein 8A, RNA binding protein 8A, RNA binding protein Y14, Y14, ZNRP, ZRNP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295284, 60365, 9939 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q27W01, Q9CWZ3, Q9Y5S9 | |
Rbm8a | |
RBM8A,Y14 Fusion Protein Ag6800 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |