Learn More
Thermo Scientific™ Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers
Maximize space using Thermo Scientific™ Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers.
£141.00 - £9905.00
Products 14
Product Code Brand Description For Use With (Equipment) Holds Price Quantity & Availability  
Product Code Brand Description For Use With (Equipment) Holds Price Quantity & Availability  
11369311
View Documents
Thermo Scientific™
398321
Flexible Side Access Microplate Rack w/Locking Rods 13 cu. ft. capacity upright freezers Microplates with Lids
£170.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
11389311
View Documents
Thermo Scientific™
398323
Flexible Side Access Microplate Rack w/Locking Rods 17.3 and 23 cu. ft. capacity upright freezers Microplates with Lids
£176.81
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
11399311
View Documents
Thermo Scientific™
398324
Adjustable Side Access Rack 13 cu. ft. capacity upright freezers 2 in. Boxes (12)
£149.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 11-08-2021
Add to basket
11309321
View Documents
Thermo Scientific™
398325
Side Access Rack 13 cu. ft. capacity upright freezers pk
£154.07
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
11319321
View Documents
Thermo Scientific™
398326
Adjustable Side Access Rack 17.3 and 23 cu. ft. capacity upright freezers 2 in. Boxes (16)
£175.78
Each
View alternative products
Save Up to 
»
Estimated Shipment: 11-08-2021
Add to basket
11329321
View Documents
Thermo Scientific™
398327
Side Access Rack 17.3 and 23 cu. ft. capacity upright freezers 2 in. Boxes (12)
£175.78
Each
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Add to basket
11359321
View Documents
Thermo Scientific™
398330
Sliding drawer rack 17.3 and 23 cu. ft. capacity upright freezers 3 in. Boxes (12)
£163.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2021
Add to basket
11369321
View Documents
Thermo Scientific™
398331
Sliding Drawer Rack 17.3 and 23 cu. ft. capacity upright freezers 2 in. Boxes (16)
£163.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2021
Add to basket
11379321
View Documents
Thermo Scientific™
398332
Sliding Drawer Rack 13 cu. ft. capacity upright freezers 2 in. Boxes (12)
£141.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2021
Add to basket
11389321
View Documents
Thermo Scientific™
398333
Side Access Microplate Rack 17.3, 23 and 28.8 cu. ft. capacity upright freezers Microplates
£175.78
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
11399321
View Documents
Thermo Scientific™
398334
Side Access Microplate Rack 13 cu. ft. capacity upright freezers Microplates
£159.24
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-10-2021
Add to basket
11376311
View Documents
Thermo Scientific™
398335
Side Access Deepwell Microplate Rack 17.3, 23 and 28.8 cu. ft. capacity upright freezers Microplates
£170.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
10731310
View Documents
Thermo Scientific™
920099
Sliding Drawer Rack 13 cu. ft. capacity upright freezers pk
£146.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Add to basket
13458649
View Documents
Thermo Scientific™
ULT320
Sliding Drawer Rack 17.3 and 23 cu. ft. capacity upright freezers 2 in. Boxes (16) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ ;Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details