missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminohexane, ChiPros 99+%, ee 96+%, Thermo Scientific Chemicals

£33.90 - £413.00

Chemical Identifiers

CAS 70095-40-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00671627
InChI Key WGBBUURBHXLGFM-ZCFIWIBFSA-N
Synonym r-2-aminohexane, 2r-hexan-2-amine, r-2-hexylamine, 2-hexanamine, r, r-2-hexaneamine, pubchem6753, 2r-2-hexanamine, r-hexan-2-amine, 2-hexanamine, 2r, r-+-2-aminohexane
PubChem CID 6993464
IUPAC Name (2R)-hexan-2-amine
SMILES CCCCC(C)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314206
View Documents
Thermo Scientific Alfa Aesar
L19304.03
1 g
£33.90
1g
Please to purchase this item. Need a web account? Register with us today!
11324206
View Documents
Thermo Scientific Alfa Aesar
L19304.06
5 g
£117.00
5g
Please to purchase this item. Need a web account? Register with us today!
11334206
View Documents
Thermo Scientific Alfa Aesar
L19304.14
25 g
£413.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70095-40-8
101.193
WGBBUURBHXLGFM-ZCFIWIBFSA-N
6993464
CCCCC(C)N
C6H15N
MFCD00671627
r-2-aminohexane, 2r-hexan-2-amine, r-2-hexylamine, 2-hexanamine, r, r-2-hexaneamine, pubchem6753, 2r-2-hexanamine, r-hexan-2-amine, 2-hexanamine, 2r, r-+-2-aminohexane
(2R)-hexan-2-amine

Specifications

70095-40-8
116°C
C6H15N
UN2733
r-2-aminohexane, 2r-hexan-2-amine, r-2-hexylamine, 2-hexanamine, r, r-2-hexaneamine, pubchem6753, 2r-2-hexanamine, r-hexan-2-amine, 2-hexanamine, 2r, r-+-2-aminohexane
CCCCC(C)N
101.193
101.19
ee ≥96%
<-20°C
−3°C (26°F)
MFCD00671627
1718994
WGBBUURBHXLGFM-ZCFIWIBFSA-N
(2R)-hexan-2-amine
6993464
≥99%
(R)-(-)-2-Aminohexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

SDS
Documents

Documents

Certificates

RUO – Research Use Only