Learn More
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Brand:  Thermo Scientific™ R4717016
Product Code. 12936037
Description
Description
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control proceduresbullets1 (end)
Includes:
10 preserved organism vials and 10 rehydration vials.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6538TM*
Quanti-Cult Plus
Cocci
100 Test/Kit
FDA 209
Staphylococcus aureus subsp. aureus ATCC™ 6538™
Quality Control Organism
Quantitative QC
100 Tests/Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538™ > Staphylococcus aureus subspecies aureus ATCC 6538
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.