Learn More
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals
£29.55 - £161.81
Chemical Identifiers
CAS | 123-75-1 |
---|---|
Molecular Formula | C4H10ClN |
Molecular Weight (g/mol) | 107.58 |
MDL Number | MFCD00005249 |
InChI Key | FCLZCOCSZQNREK-UHFFFAOYSA-N |
Synonym | tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine |
PubChem CID | 31268 |
ChEBI | CHEBI:33135 |
IUPAC Name | hydrogen pyrrolidine chloride |
SMILES | [H+].[Cl-].C1CCNC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11976761
|
Thermo Scientific Acros
446141000 |
100 mL | AcroSeal™ Glass bottle |
£29.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996761
|
Thermo Scientific Acros
446148000 |
800 mL | AcroSeal™ Glass bottle |
£161.81
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-75-1 | |
107.58 | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
31268 | |
hydrogen pyrrolidine chloride |
C4H10ClN | |
MFCD00005249 | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
CHEBI:33135 | |
[H+].[Cl-].C1CCNC1 |
Specifications
123-75-1 | |
0.8660g/mL | |
3°C | |
C4H10ClN | |
MFCD00005249 | |
0.866 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents | |
[H+].[Cl-].C1CCNC1 | |
107.58 | |
CHEBI:33135 | |
71.11 | |
Pyrrolidine |
-63.0°C | |
86.0°C to 88.0°C | |
99.4% min. (GC) | |
1.4421 to 1.4441 | |
15,8129 | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
hydrogen pyrrolidine chloride | |
31268 | |
0.94 mPa.s (20°C) | |
99.5+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-648-7
- Pyrrolidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Pyrrolidine
RUO – Research Use Only