Learn More
Cole-Parmer™ PVDF Manifold, 4 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Brand:  Cole-Parmer™ 31523-39
 View more versions of this product
Product Code. 11775278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Specifications
Manifold
4 1/8 in.
-65°F
3/8 in. x 1/4 in.
1/4 in. NPT(F)
1.25 in.
1.25 in.
230°F
PVDF
10.48 cm
-54°C
3/8 in. NPT(F)
3.18 cm
3.18 cm
110 psi
110°C